Skip to Content
Merck
All Photos(3)

Key Documents

737461

Sigma-Aldrich

1H,1H,2H,2H-Perfluorooctanephosphonic acid

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$252.00
5 G
$877.00

$252.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$252.00
5 G
$877.00

About This Item

Empirical Formula (Hill Notation):
C8H6F13O3P
CAS Number:
Molecular Weight:
428.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$252.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

95%

form

solid

mp

168-173 °C

storage temp.

2-8°C

SMILES string

OP(O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8H6F13O3P/c9-3(10,1-2-25(22,23)24)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1-2H2,(H2,22,23,24)

InChI key

DEXIXSRZQUFPIK-UHFFFAOYSA-N

General description

1H,1H,2H,2H-Perfluorooctanephosphonic (PFOPA) acid is a fluoroalkyl phosphonic acid that contains 8 fluorinated carbon atoms. It forms a self-assembled layer on the substrate due to the adhesion of the phosphonates. The surface adhesion is a component of the acid base linkage that is enhanced by the hydroxyl groups.[1][2]

Application

PFOPA can functionalize gallium nitride (GaN) to tune the optical properties, which can potentially be used in chemical sensor based applications.[3] It can modify the surface characteristics of copper substrates that find usage in printed circuit boards as copper foils.[2] PFOPA can also be coated on the indium tin oxide substrate, which may be utilized in organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs).[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified surface chemistry, potential, and optical properties of polar gallium nitride via long chained phosphonic acids.
Wilkins SJ, et al.
Applied Surface Science, 327(38), 498-503 (2015)
Phosphonic acid modification of indium- tin oxide electrodes: Combined XPS/UPS/contact angle studies.
Paniagua SA, et al.
The Journal of Physical Chemistry C, 112(21), 7809-7817 (2008)
Direct Patterning of Self-Assembled Monolayers by Stamp Printing Method and Applications in High Performance Organic Field-Effect Transistors and Complementary Inverters.
Zhang Z, et al.
Advances in Functional Materials, 25(38), 6112-6121 (2015)
Advanced surface modification of indium tin oxide for improved charge injection in organic devices.
Hanson EL, et al.
Journal of the American Chemical Society, 127(28), 10058-10062 (2005)
Surface characterization of copper substrates modified with carboxyl terminated phosphonic acids.
Nothdurft P, et al.
International Journal of Adhesion and Adhesives, 84(38), 143-152 (2018)

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service