Skip to Content
Merck
All Photos(1)

Key Documents

737410

Sigma-Aldrich

1,10-Decyldiphosphonic acid

>97%

Synonym(s):

1,10-Decanediphosphonic acid, Decyl-1,10-diphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24O6P2
CAS Number:
Molecular Weight:
302.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

>97%

form

solid

mp

198-203 °C

SMILES string

OP(O)(=O)CCCCCCCCCCP(O)(O)=O

InChI

1S/C10H24O6P2/c11-17(12,13)9-7-5-3-1-2-4-6-8-10-18(14,15)16/h1-10H2,(H2,11,12,13)(H2,14,15,16)

InChI key

AYQDAYSTSMCOCJ-UHFFFAOYSA-N

Application

Used in the modification of oxide surfaces and also in the synthesis of nanotube templates.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifeng Hou et al.
Journal of the American Chemical Society, 126(18), 5674-5675 (2004-05-06)
Electroless deposition of gold on the pore walls of polycarbonate templates is currently the best known method for controlling inside diameters of template-synthesized nanotubes. It would be very useful to have alternative template-based synthetic chemistries that yield nanotubes composed of
Cheon; Ik Chan; Lee; Myung Soon; Shim, et al
Bull. Korean Chem. Soc., 10, 1535-1535 (2003)

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service