Skip to Content
Merck
All Photos(1)

Key Documents

737232

Sigma-Aldrich

2-Methyl-5-(trifluoromethyl)pyrazine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F3N2
CAS Number:
Molecular Weight:
162.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

mp

38-43 °C

functional group

fluoro

SMILES string

Cc1cnc(cn1)C(F)(F)F

InChI

1S/C6H5F3N2/c1-4-2-11-5(3-10-4)6(7,8)9/h2-3H,1H3

InChI key

XZOKPWSKJRCLQO-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

120.0 °F

Flash Point(C)

48.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service