Skip to Content
Merck
All Photos(1)

Documents

736872

Sigma-Aldrich

3,4-Pentadien-1-ol

Synonym(s):

5-Hydroxy-1,2-pentadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O
CAS Number:
Molecular Weight:
84.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

refractive index

n20/D 1.476

density

0.894 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCC=C=C

InChI

1S/C5H8O/c1-2-3-4-5-6/h3,6H,1,4-5H2

InChI key

WHQNQGMPXBYFSM-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F

Flash Point(C)

48.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service