Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
736007
Vanadium(V) oxytriisopropoxide
packaged for use in deposition systems
Synonym(s):
Triisopropoxyvanadium(V) oxide, VTIP, Vanadium(V) trisisopropoxide oxide
About This Item
Recommended Products
Quality Level
form
liquid
composition
V, 20.3-21.4% titration (, by potassium permanganate)
reaction suitability
core: vanadium
refractive index
n20/D 1.479 (lit.)
bp
80-82 °C/2 mmHg (lit.)
density
1.035 g/mL at 25 °C (lit.)
SMILES string
CC(C)O[V](=O)(OC(C)C)OC(C)C
InChI
1S/3C3H7O.O.V/c3*1-3(2)4;;/h3*3H,1-2H3;;/q3*-1;;+3
InChI key
DSGGJXAUQHKOGQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
113.0 °F - closed cup
Flash Point(C)
45 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service