Skip to Content
Merck
All Photos(1)

Key Documents

735949

Sigma-Aldrich

Gadolinium(III) chloride

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Gadolinium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
beads

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

beads

particle size

−10 mesh

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Gadolinium Chloride is a commonly used precursor for the formation of MRI contrast agent and can be used as a Diels-Alder catalyst.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu, J.; et al.
Journal of the American Chemical Society, 7, 7245-7246 (1995)
Lillian Zhang et al.
The Journal of toxicological sciences, 37(2), 447-453 (2012-04-03)
Liver toxicity is a side effect observed with some herbal treatments, including Piper methysticum. The possible mechanisms responsible include inflammation subsequent to activation of liver macrophages and oxidative damage. Hepatotoxicity of the pharmacologically active component of Piper methysticum (kavalactones) was
Puja Gulati et al.
Naunyn-Schmiedeberg's archives of pharmacology, 386(3), 255-264 (2012-12-12)
The present study was designed to investigate the potential of gadolinium, a stretch-activated calcium channel blocker in ischemic reperfusion (I/R)-induced brain injury in mice. Bilateral carotid artery occlusion of 12 min followed by reperfusion for 24 h was given to induce cerebral
Minako Hara et al.
American journal of physiology. Cell physiology, 302(12), C1741-C1750 (2012-03-31)
When skeletal muscle is stretched or injured, satellite cells, resident myogenic stem cells positioned beneath the basal lamina of mature muscle fibers, are activated to enter the cell cycle. This signaling pathway is a cascade of events including calcium-calmodulin formation
T Hasezaki et al.
Die Pharmazie, 66(9), 698-703 (2011-10-27)
Nanomaterials have potential toxicity that is not found in micromaterials, and it is therefore essential to understand their biological activity and potential toxicity. We focused on silica nanoparticles, since it was previously reported that the intravenous administration of silica nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service