Skip to Content
Merck
All Photos(1)

Key Documents

735604

Sigma-Aldrich

1-Ethylcyclopentanol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
96%

Assay

96%

form

liquid

refractive index

n20/D 1.454

density

0.909 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCC1(O)CCCC1

InChI

1S/C7H14O/c1-2-7(8)5-3-4-6-7/h8H,2-6H2,1H3

InChI key

LPCWIFPJLFCXRS-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.9 °F

Flash Point(C)

49.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service