Skip to Content
Merck
All Photos(1)

Key Documents

732265

Sigma-Aldrich

1-Boc-3-(aminomethyl)azetidine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$189.00

$189.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$189.00

About This Item

Empirical Formula (Hill Notation):
C9H18N2O2
Molecular Weight:
186.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00


Estimated to ship onApril 22, 2025


Request a Bulk Order

form

liquid

Quality Level

refractive index

n/D 1.4691

density

1.0234 g/mL at 25 °C

functional group

amine

SMILES string

CC(C)(C)OC(=O)N1CC(CN)C1

InChI

1S/C9H18N2O2/c1-9(2,3)13-8(12)11-5-7(4-10)6-11/h7H,4-6,10H2,1-3H3

InChI key

XSJPKMUFBHSIRA-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service