Skip to Content
Merck
All Photos(1)

Key Documents

730203

Sigma-Aldrich

Methyl pyridazine-4-carboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$274.00

$274.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$274.00

About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$274.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

63-69 °C

functional group

ester

SMILES string

COC(=O)c1ccnnc1

InChI

1S/C6H6N2O2/c1-10-6(9)5-2-3-7-8-4-5/h2-4H,1H3

InChI key

IJFLWNYKGMQQOW-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yutao Jiang et al.
Journal of chromatography. A, 1411, 63-68 (2015-08-11)
In this paper, we introduce a high throughput LCMS/UV/CAD/CLND system that improves upon previously reported systems by increasing both the quantitation accuracy and the range of compounds amenable to testing, in particular, low molecular weight "fragment" compounds. This system consists

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service