Skip to Content
Merck
All Photos(1)

Key Documents

729817

Sigma-Aldrich

2-Chloroethyl acrylate

contains >100 ppm MEHQ as inhibitor, 97%

Synonym(s):

CEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2O(CO)CHCH2
CAS Number:
Molecular Weight:
134.56
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

contains

>100 ppm MEHQ as inhibitor

refractive index

n20/D 1.125

density

1.125 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClCCOC(=O)C=C

InChI

1S/C5H7ClO2/c1-2-5(7)8-4-3-6/h2H,1,3-4H2

InChI key

WHBAYNMEIXUTJV-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.9 °F

Flash Point(C)

59.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reyhaneh Keshmiri et al.
Medicine, 98(23), e15841-e15841 (2019-06-07)
The aim of the study was to determine whether exclusive breastfeeding or exclusive formula feeding is more cost-effective when a Canadian mother with HIV is adherent to antiretroviral therapy and has full virologic suppression. Current Canadian guidelines recommend that mothers
Shijun Chen et al.
Clinica chimica acta; international journal of clinical chemistry, 466, 167-171 (2017-01-29)
Circular RNAs (circRNA) are an abundant class of non-coding RNAs in mammalian cells. However, their value in the diagnosis of cancers remains unknown. In this study, we focused on hsa_circ_0000190, which was found to be down-regulated in gastric cancer tissues
A M Sirunyan et al.
Physical review letters, 120(20), 202005-202005 (2018-06-05)
A search for resonancelike structures in the B_{s}^{0}π^{±} invariant mass spectrum is performed using proton-proton collision data collected by the CMS experiment at the LHC at sqrt[s]=8  TeV, corresponding to an integrated luminosity of 19.7  fb^{-1}. The B_{s}^{0} mesons are reconstructed in
Tawfiq Nasr Allah et al.
Angewandte Chemie (International ed. in English), 58(32), 10884-10887 (2019-06-01)
The first iron catalysts able to promote the formal insertion of CO into the C-N bond of amines are reported. Using low-valent iron complexes, including K2 [Fe(CO)4 ], amides are formed from aromatic and aliphatic amines, in the presence of
Yi Shen et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 28(7), 1212-1220 (2019-04-17)
Circulating miRNAs have been identified as diagnostic biomarkers for esophageal squamous cell carcinoma (ESCC), but their efficacy in discovering early-stage ESCC is still unsatisfying. Esophageal squamous dysplasia (ESD) is the precursor lesion of ESCC. Notably, little is known about the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service