Skip to Content
Merck
All Photos(1)

Key Documents

729426

Sigma-Aldrich

5-Cyano-2-methoxybenzaldehyde

97%

Synonym(s):

3-Formyl-4-methoxybenzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$119.00

$119.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$119.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

115-120 °C

functional group

aldehyde
nitrile

storage temp.

2-8°C

SMILES string

COc1ccc(cc1C=O)C#N

InChI

1S/C9H7NO2/c1-12-9-3-2-7(5-10)4-8(9)6-11/h2-4,6H,1H3

InChI key

HJDXZGCTGAWUFZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service