Skip to Content
Merck
All Photos(1)

Key Documents

728993

Sigma-Aldrich

2-Naphthyl propiolate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

65-72 °C

functional group

ester

SMILES string

O=C(Oc1ccc2ccccc2c1)C#C

InChI

1S/C13H8O2/c1-2-13(14)15-12-8-7-10-5-3-4-6-11(10)9-12/h1,3-9H

InChI key

ANECZRYIRALULR-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service