Skip to Content
Merck
All Photos(1)

Key Documents

728209

Sigma-Aldrich

(3-Bromo-1-propynyl)benzene solution

~75% in toluene

Synonym(s):

3-Bromo-1-phenylpropyne

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$530.00

$530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C9H7Br
CAS Number:
Molecular Weight:
195.06
Beilstein:
878435
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solution

concentration

~75% in toluene

storage temp.

−20°C

SMILES string

BrCC#Cc1ccccc1

InChI

1S/C9H7Br/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,8H2

InChI key

RKNCIBMWPVZEAJ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service