Skip to Content
Merck
All Photos(1)

Key Documents

727601

Sigma-Aldrich

2,2′-Biphenyldiamine

≥96.5% (GC)

Synonym(s):

2,2′-Diaminobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
1949579
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

≥96.5% (GC)

form

solid

SMILES string

Nc1ccccc1-c2ccccc2N

InChI

1S/C12H12N2/c13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H,13-14H2

InChI key

HOLGXWDGCVTMTB-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service