Skip to Content
Merck
All Photos(1)

Key Documents

727067

Sigma-Aldrich

(R)-(−)-2-Chloromandelic acid

ChiPros®, produced by BASF, 98%, ≥97.5% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(OH)CO2H
CAS Number:
Molecular Weight:
186.59
Beilstein:
2693370
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Assay

≥97.5% (HPLC)
98%

form

crystals

optical purity

enantiomeric excess: ≥98.5%

mp

119-121 °C (lit.)

SMILES string

O[C@@H](C(O)=O)c1ccccc1Cl

InChI

1S/C8H7ClO3/c9-6-4-2-1-3-5(6)7(10)8(11)12/h1-4,7,10H,(H,11,12)/t7-/m1/s1

InChI key

RWOLDZZTBNYTMS-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service