Skip to Content
Merck
All Photos(1)

Key Documents

725919

Sigma-Aldrich

4,4′-Bis(bromomethyl)biphenyl

≥97.0% (HPLC)

Synonym(s):

α,α′-Dibromo-p,p′-bitolyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.00
5 G
$257.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$127.00
5 G
$257.00

About This Item

Linear Formula:
BrCH2C6H4C6H4CH2Br
CAS Number:
Molecular Weight:
340.05
Beilstein:
2444585
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (HPLC)

form

powder

mp

170-173 °C

functional group

bromo

SMILES string

BrCc1ccc(cc1)-c2ccc(CBr)cc2

InChI

1S/C14H12Br2/c15-9-11-1-5-13(6-2-11)14-7-3-12(10-16)4-8-14/h1-8H,9-10H2

InChI key

HMUGRILXVBKBID-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service