Skip to Content
Merck
All Photos(1)

Key Documents

72565

Sigma-Aldrich

Nitrilotriacetic acid trisodium salt monohydrate

≥98.0% (KT)

Synonym(s):

Sodium nitrilotriacetate tribasic monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$46.30
1 KG
$153.00

$46.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$46.30
1 KG
$153.00

About This Item

Linear Formula:
N(CH2COONa)3 · H2O
CAS Number:
Molecular Weight:
275.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$46.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98.0% (KT)

form

powder

mp

>300 °C (lit.)

functional group

amine

SMILES string

O.[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O

InChI

1S/C6H9NO6.3Na.H2O/c8-4(9)1-7(2-5(10)11)3-6(12)13;;;;/h1-3H2,(H,8,9)(H,10,11)(H,12,13);;;;1H2/q;3*+1;/p-3

InChI key

HERBOKBJKVUALN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Nitrilotriacetic acid trisodium salt monohydrate [(N(CH2CO2Na)3 H2O)] [NTA] has been used in the preparation of chemical bath deposition (CBD) solution for the deposition of quantum dots (QDs) on photoanodic substrate.[1]

It can also be can be used:
  • As a complexing agent in an electrolytic bath solution for the synthesis of magnetic materials by electrochemical method.[2]
  • As a reducing agent and colloid stabilizer for the preparation of uniform gold nanoparticles.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura T Rea et al.
Environmental science. Processes & impacts, 21(1), 89-103 (2018-11-10)
Trace metal ion speciation in natural waters is often under kinetic control due to slow exchange reactions involving multidentate ligands (both natural and anthropogenic) and constituent ions (e.g. calcium). Incomplete understanding of the kinetic behavior (rates, rate laws, and mechanisms)
Charge recombination control for high efficiency CdS/CdSe quantum dot co-sensitized solar cells with multi-ZnS layers
Wu Q, et al.
Dalton Transactions, 47(7), 2214-2221 (2018)
Amal Al Teneiji et al.
Molecular genetics and metabolism, 120(3), 235-242 (2017-01-27)
Congenital disorders of glycosylation (CDG) are inborn defects of glycan metabolism. They are multisystem disorders. Analysis of transferrin isoforms is applied as a screening test for CDG type I (CDG-I) and type II (CDG-II). We performed a retrospective cohort study
Electrochemical synthesis of Fe-W and Fe-WP magnetic amorphous films and Fe-W nanowires
Barbano, EP and Carlos, IA and Vall
Surface and Coatings Technology, 324(7), 80-84 (2017)
Nitrilotriacetic acid: A novel reducing agent for synthesizing colloidal gold
Njagi JI and Goia DV
Journal of Colloid and Interface Science, 421(7), 27-32 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service