Skip to Content
Merck
All Photos(1)

Key Documents

725315

Sigma-Aldrich

(S)-2-(3-Chloro-2,2-dimethyl-propionylamino)-3-methylbutanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20ClNO2
Molecular Weight:
221.72
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

73-79 °C

SMILES string

CC(C)[C@@H](CO)NC(=O)C(C)(C)CCl

InChI

1S/C10H20ClNO2/c1-7(2)8(5-13)12-9(14)10(3,4)6-11/h7-8,13H,5-6H2,1-4H3,(H,12,14)/t8-/m1/s1

InChI key

NBXJRNNARUAEFQ-MRVPVSSYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service