Skip to Content
Merck
All Photos(1)

Key Documents

724467

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-methacrylic acid)

methacrylic acid 5 mol %, average Mn 50,000

Synonym(s):

Poly(NIPAM-co-MAA), Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)m (C4H6O2)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn 50,000

composition

methacrylic acid, 5 mol %

mp

>300 °C

InChI

1S/C6H11NO.C4H6O2/c1-4-6(8)7-5(2)3;1-3(2)4(5)6/h4-5H,1H2,2-3H3,(H,7,8);1H2,2H3,(H,5,6)

InChI key

BGJOTKHBFYMJST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intelligent Swelling/Collapsing copolymer that can be used as a temperature- and pH-sensitive materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Zhang et al.
The Journal of pharmacy and pharmacology, 56(5), 611-620 (2004-05-15)
A new glucose-responsive polymeric composite membrane that provided pulsatile insulin release was developed in our laboratory previously. To develop a clinically useful insulin delivery system, this study was designed to investigate factors influencing insulin stability during delivery by this membrane.
Ricardo G Sousa et al.
Journal of controlled release : official journal of the Controlled Release Society, 102(3), 595-606 (2005-02-01)
The loading of an antihypertensive cationic drug, diltiazem hydrochloride (DIL.HCl), into poly(N-isopropylacrylamide) [P(N-iPAAm)], poly(methacrylic acid) [P(MAA)], and their poly[(N-isopropylacrylamide)-co-(methacrylic acid)] P[(N-iPAAm)-co-(MAA)] hydrogels as well as their release behaviour have been investigated. For this purpose, two series of hydrogels have been
Kai Zhang et al.
Biomacromolecules, 5(4), 1248-1255 (2004-07-13)
To elucidate the mechanism of stimuli-responsive permeability and to optimize the design, the nanostructure of polymeric composite membranes, developed in our laboratory, was characterized. The membranes were prepared to contain various amounts of stimuli-responsive nanoparticles of poly(N-isopropylacrylamide-co-methacrylic acid), with or
Yunlu Dai et al.
ACS nano, 6(4), 3327-3338 (2012-03-23)
In this study, we report a new controlled release system based on up-conversion luminescent microspheres of NaYF(4):Yb(3+)/Er(3+) coated with the smart hydrogel poly[(N-isopropylacrylamide)-co-(methacrylic acid)] (P(NIPAM-co-MAA)) (prepared using 5 mol % of MAA) shell. The hybrid microspheres show bright up-conversion fluorescence
Pierre Simard et al.
International journal of pharmaceutics, 381(2), 86-96 (2009-05-19)
A promising avenue in cancer therapy using liposomal formulations is the combination of site-specific delivery with triggered drug release. The use of trigger mechanisms in liposomes could be relevant for drugs susceptible to lysosomal hydrolytic/enzymatic degradation. Here, we propose a

Articles

Poly(N-isopropylacrylamide), or PNIPAM, is a stimuli-responsive polymer that responds to changes in pH and temperature and has a LCST around 32 C.

Tissue engineering has become a key therapeutic tool in the treatment of damaged or diseased organs and tissues, such as blood vessels and urinary bladders.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service