Skip to Content
Merck
All Photos(1)

Key Documents

721131

Sigma-Aldrich

(4-Benzyloxyphenyl)tributylstannane

Synonym(s):

[4-(Benzyloxy)phenyl]tributyltin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38OSn
CAS Number:
Molecular Weight:
473.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.546

density

1.128 g/cm3 at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccc(OCc2ccccc2)cc1

InChI

1S/C13H11O.3C4H9.Sn/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13;3*1-3-4-2;/h1,3-10H,11H2;3*1,3-4H2,2H3;

InChI key

DRDMDSYSKAAPQQ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of epoxy-substituted aryl alkenes by Stille coupling reactions with allylic carbonates
  • Preparation of polycyclic meroterpenoids based on Stille couplings and titanocene catalysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service