Skip to Content
Merck
All Photos(1)

Documents

721018

Supelco

Ethylbenzene solution

NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

concentration

0.1% in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

1H sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rezazadeh Azari et al.
The international journal of occupational and environmental medicine, 3(1), 39-44 (2012-10-02)
Benzene, toluene, ethylbenzene and xylene (BTEX) are the most important toxic volatile compounds in the air and could be easily absorbed through the respiratory tract. In recent years, the risk of exposure to BTEX compounds, especially benzene as a carcinogen
You-Ya Zhou et al.
Environmental monitoring and assessment, 185(4), 3037-3048 (2012-09-11)
A method based on headspace (HS) sampling coupling with portable gas chromatography (GC) with photo ionization detector (PID) was developed for rapid determination of benzene, toluene, ethylbenzene, and xylenes (BTEX) in soils. Optimal conditions for HS gas sampling procedure were
Ch Vlachokostas et al.
Environmental pollution (Barking, Essex : 1987), 177, 125-134 (2013-03-19)
Emission from road traffic has become the most important source of local air pollution in numerous European cities. Epidemiological research community has established consistent associations between traffic-related substances and various health outcomes. Nevertheless, the vast majority of urban areas are
Lili Zhang et al.
Chemosphere, 90(4), 1340-1347 (2012-09-11)
A new strain Mycobacterium cosmeticum byf-4 able to simultaneously degrade benzene, toluene, ethylbenzene, and o-xylene (BTE(o-)X) compounds has been isolated and identified previously in our laboratory. We further report here the extent of degradation of every BTE(o-)X component, and unravel
Carla G A da Silva et al.
Journal of chromatography. A, 1260, 81-87 (2012-09-12)
Endcapped stationary phases were prepared after thermal immobilization of poly(methyloctadecylsiloxane) (PMODS) onto zirconized and titanized silica supports. These new stationary phases have lower densities of residual hydroxyl groups, according to infrared spectroscopy and 29Si CP-MAS NMR and as shown by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service