Skip to Content
Merck
All Photos(2)

Key Documents

720968

Sigma-Aldrich

Biphenyl-3,3′,5,5′-tetracarboxylic acid

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O8
CAS Number:
Molecular Weight:
330.25
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>400 °C

greener alternative category

SMILES string

OC(=O)c1cc(cc(c1)-c2cc(cc(c2)C(O)=O)C(O)=O)C(O)=O

InChI

1S/C16H10O8/c17-13(18)9-1-7(2-10(5-9)14(19)20)8-3-11(15(21)22)6-12(4-8)16(23)24/h1-6H,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

QURGMSIQFRADOZ-UHFFFAOYSA-N

General description

Biphenyl-3,3′,5,5′-tetracarboxylic acid (H4BPTC) is a carboxylic ligand material, which has multiple co-ordinated sites, and the carboxyl groups lie in the position of 120° that extend in all the directions. It has a luminescent property with an emission band maximum at 405 nm.[1][2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Biphenyl-3,3′,5,5′-tetracarboxylic acid is used for fabricating CPF-1,Cu2(bptc)(H2O)2,MOF-501,MOF-502,MOF-505, NOTT-100, NOTT-400, NOTT-401, SNU-M11,SNU-M10 a highly symmetrical chiral Mg-based metal organic framework (MOF) with nanotubular channels. These are used for selective gas adsorption.[3][4][5][6][7]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly porous and robust scandium-based metal?organic frameworks for hydrogen storage.
Ibarra I A, et al.
Chemical Communications (Cambridge, England), 47(29), 8304-8306 (2011)
A new PtS-type heterometallic compound constructed from biphenyl-3, 3?, 5, 5?-tetracarboxylate: Synthesis, structure and luminescent property
Zhu L, et al.
Journal of Molecular Structure, 1067, 239-242 (2014)
High H2 adsorption by coordination?framework materials.
Lin X, et al.
Angewandte Chemie (International Edition in English), 45(44), 7358-7364 (2006)
Transformation of a metal? organic framework from the NbO to PtS net.
Chen B, et al.
Inorganic Chemistry, 44(2), 181-183 (2005)
Two new Cd (II) coordination polymer based on Biphenyl-3, 3?, 5, 5?-tetracarboxylic acid
Tan J, et al.
Inorganic Chemistry Communications, 87, 36-39 (2018)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service