Skip to Content
Merck
All Photos(1)

Key Documents

72078

Sigma-Aldrich

(−)-α-Neoclovene

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(1S,7aS)-1,2,3,6,7,7a-Hexahydro-2,2,4,7a-tetramethyl-1,3a-ethano-3aH-indene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −79±2°, neat

refractive index

n20/D 1.508

density

0.951 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1=CCC[C@@]2(C)C3CC[C@@]12CC3(C)C

InChI

1S/C15H24/c1-11-6-5-8-14(4)12-7-9-15(11,14)10-13(12,2)3/h6,12H,5,7-10H2,1-4H3/t12?,14-,15+/m0/s1

InChI key

ZCJQJJWNFDNQGZ-JQXSQYPDSA-N

Other Notes

Rearrangement product of caryophyllene[1]; Main volatile constituent of ginseng[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Parker et al.
Journal of the Chemical Society, 2634-2634 (1969)
K. Yoshihara et al.
Bulletin of the Chemical Society of Japan, 48, 2078-2078 (1975)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service