Skip to Content
Merck
All Photos(1)

Key Documents

720070

Sigma-Aldrich

Palladium(II) acetate, recrystallized

97%

Synonym(s):

Pd(OAc)2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$222.00

$222.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$222.00

About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$222.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

227-231 °C

SMILES string

CC(=O)O[Pd]OC(C)=O

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

General description

Palladium(II)acetate, recrystallized, is commonly used as a catalyst in cross-coupling reactions. Prominent applications include the Buchwald-Hartwigamination reaction, Heck reaction, Suzuki coupling, Stille coupling, Sonogashira coupling, and Negishi coupling reactions.

Application

Palladium(II) acetate, recrystallized can be used as a catalyst:
  • In Stille cross-coupling reaction between benzylic halides and aryl tributyltin to synthesize diarylmethane derivatives in the presence of phosphine ligand.
  • For the α-arylation of ketones with aryl halides to synthesize arylmethyl ketones in the presence of n-butylbis(1-adamantyl)-phosphine ligand.
  • In Suzuki-Miyaura cross-coupling reaction between aryl bromides and aryl boronic acids to synthesize biaryl derivatives in the presence of phosphane ligand.
  • In Buchwald-Hartwig amination reaction between aryl chlorides and anilines to synthesize arylated anilines in the presence of N-heterocyclic carbene ligand.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinyue Ding et al.
Nature communications, 5, 5474-5474 (2014-11-19)
Non-aromatic nitrogen- and oxygen-containing heterocycles such as piperidines and pyrans are prevalent components of natural products and pharmaceutical drugs. Although it has been a workhorse as a synthetic method for assembling unsaturated sp(2)-hybridized substrates, transition metal-catalysed cross-coupling chemistry is traditionally

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service