Skip to Content
Merck
All Photos(1)

Key Documents

719986

Sigma-Aldrich

(1,5-Cyclooctadiene)bis(trimethylsilylmethyl)palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H34PdSi2
CAS Number:
Molecular Weight:
389.03
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: palladium

mp

52-62 °C

shipped in

dry ice

storage temp.

−20°C

SMILES string

C1CC=CCCC=C1.C[Si](C)(C)C[Pd]C[Si](C)(C)C

InChI

1S/C8H12.2C4H11Si.Pd/c1-2-4-6-8-7-5-3-1;2*1-5(2,3)4;/h1-2,7-8H,3-6H2;2*1H2,2-4H3;/b2-1-,8-7-;;;

InChI key

OOUJKOJUEUVIOB-PHFPKPIQSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service