Skip to Content
Merck
All Photos(2)

Key Documents

718009

Sigma-Aldrich

Dicyclohexylzinc solution

0.4 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22Zn
CAS Number:
Molecular Weight:
231.69
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

0.4 M in diethyl ether

density

0.756 g/mL at 25 °C

SMILES string

C1CCC(CC1)[Zn]C2CCCCC2

InChI

1S/2C6H11.Zn/c2*1-2-4-6-5-3-1;/h2*1H,2-6H2;

InChI key

NFBPCQSTZITERU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service