Skip to Content
Merck
All Photos(2)

Key Documents

716928

Sigma-Aldrich

Gold nanorods

palladium coated, 25 nm diameter, 100 μg/mL in H2O, dispersion

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$1,620.00

$1,620.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$1,620.00

About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$1,620.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

material

palladium

form

colloidal suspension
dispersion
nanorod

concentration

100 μg/mL in H2O

diam. × L

25 nm × 75 nm

diameter

25 nm

density

1 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service