Skip to Content
Merck
All Photos(1)

Key Documents

716715

Sigma-Aldrich

(S)-2-[(Diphenylphosphino)methyl]pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$112.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$112.00

About This Item

Empirical Formula (Hill Notation):
C17H20NP
CAS Number:
Molecular Weight:
269.32
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

optical activity

[α]22/D −24.0°, c = 0.5 in ethanol

refractive index

n20/D 1.614

density

1.043 g/mL at 25 °C

functional group

phosphine

SMILES string

C1CN[C@@H](C1)CP(c2ccccc2)c3ccccc3

InChI

1S/C17H20NP/c1-3-9-16(10-4-1)19(14-15-8-7-13-18-15)17-11-5-2-6-12-17/h1-6,9-12,15,18H,7-8,13-14H2/t15-/m0/s1

InChI key

IWRBGJKCDORZHK-HNNXBMFYSA-N

Application

Catalytic ligand in:
  • Ru-catalyzed regioselective lactonization of unsymmetrical 1,4-diols to give lactone lignans
  • Preparation of vinylindanecarboxaldehyde by asymmetric intramolecular allylation of propenylbenzenepropanal

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service