Skip to Content
Merck
All Photos(1)

Key Documents

716537

Sigma-Aldrich

Dimethyl 4,5-imidazoledicarboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$60.00

$60.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$60.00

About This Item

Empirical Formula (Hill Notation):
C7H8N2O4
CAS Number:
Molecular Weight:
184.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$60.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

207-212 °C

functional group

ester

SMILES string

COC(=O)c1nc[nH]c1C(=O)OC

InChI

1S/C7H8N2O4/c1-12-6(10)4-5(7(11)13-2)9-3-8-4/h3H,1-2H3,(H,8,9)

InChI key

CUIWFAXEALIQJS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service