Skip to Content
Merck
All Photos(1)

Key Documents

716458

Sigma-Aldrich

1,3-Bis(2,6-di-i-propylphenyl)imidazolidin-2-ylidene

Synonym(s):

SIPr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N2
CAS Number:
Molecular Weight:
390.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: catalyst

mp

170-175 °C

storage temp.

−20°C

SMILES string

CC(C)c1cccc(C(C)C)c1N2[C]N(CC2)c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H38N2/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8/h9-14,18-21H,15-16H2,1-8H3

InChI key

XZDYFCGKKKSOEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Transformation of organometallic iron complexes
  • Selective production of 5-hydroxymethylfurfural from glucose and fructose
  • Annulation of heterocycles via redox processing
  • Pyridines
  • Cyclotrimerization of isocyanates
  • Ring-opening alkylation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service