Skip to Content
Merck
All Photos(1)

Key Documents

716286

Sigma-Aldrich

3-(Dimethylamino)-2-iodoacrolein

97%

Synonym(s):

2-Iodo-3-(dimethylamino)acrylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8INO
CAS Number:
Molecular Weight:
225.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

73-77 °C

storage temp.

−20°C

SMILES string

CN(C)\C=C(/I)C=O

InChI

1S/C5H8INO/c1-7(2)3-5(6)4-8/h3-4H,1-2H3/b5-3-

InChI key

AHWCNVWCBYBGQI-HYXAFXHYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service