Skip to Content
Merck
All Photos(2)

Key Documents

716243

Sigma-Aldrich

1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester

97%

Synonym(s):

1-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$179.00

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$179.00

About This Item

Empirical Formula (Hill Notation):
C12H21BN2O2
CAS Number:
Molecular Weight:
236.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

36-44 °C

storage temp.

2-8°C

SMILES string

CC(C)n1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C12H21BN2O2/c1-9(2)15-8-10(7-14-15)13-16-11(3,4)12(5,6)17-13/h7-9H,1-6H3

InChI key

OGYYMVGDKVJYSU-UHFFFAOYSA-N

Application

1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester can be used as a reactant to synthesize:
  • Imidazo[1,2-a]pyrazine based phosphodiesterase 10A inhibitors.[1]
  • Pyrido[3,4-d]pyrimidin-4-(3H)-one based KDM4 and KDM5 (histone lysine demethylase families) inhibitors.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell penetrant inhibitors of the KDM4 and KDM5 families of histone lysine demethylases. 2. Pyrido [3, 4-d] pyrimidin-4 (3 H)-one derivatives
Westaway SM, et al.
Journal of Medicinal Chemistry, 59(4), 1370-1387 (2016)
Discovery of a potent, selective, and orally active phosphodiesterase 10A inhibitor for the potential treatment of schizophrenia
Bartolome?-Nebreda JM, et al.
Journal of Medicinal Chemistry, 57(10), 4196-4212 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service