Skip to Content
Merck
All Photos(1)

Key Documents

716022

Sigma-Aldrich

2-Vinylpyrazine

97%

Synonym(s):

2-Ethenylpyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$135.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$135.00

About This Item

Empirical Formula (Hill Notation):
C6H6N2
CAS Number:
Molecular Weight:
106.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.560

density

1.041 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=Cc1cnccn1

InChI

1S/C6H6N2/c1-2-6-5-7-3-4-8-6/h2-5H,1H2

InChI key

KANZWHBYRHQMKZ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service