Skip to Content
Merck
All Photos(1)

Key Documents

715689

Sigma-Aldrich

3-Furoyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$411.00

$411.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$411.00

About This Item

Empirical Formula (Hill Notation):
C5H3ClO2
CAS Number:
Molecular Weight:
130.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$411.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

refractive index

n20/D 1.501

mp

25-30 °C

functional group

acyl chloride

SMILES string

ClC(=O)c1ccoc1

InChI

1S/C5H3ClO2/c6-5(7)4-1-2-8-3-4/h1-3H

InChI key

BTUIFMCWPFMNRG-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

136.9 °F

Flash Point(C)

58.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service