Skip to Content
Merck
All Photos(1)

Key Documents

715239

Sigma-Aldrich

N-Boc-3,4-dihydro-2H-pyridine

97%

Synonym(s):

1-N-Boc-1,2,3,4-tetrahydropyridine, 3,4-Dihydro-1(2H)-pyridinecarboxylic acid 1,1-dimethylethyl ester, N-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydropyridine, tert-Butyl 3,4-dihydropyridine-1(2H)-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$204.00

About This Item

Empirical Formula (Hill Notation):
C10H17NO2
CAS Number:
Molecular Weight:
183.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.477

density

0.988 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CCCC=C1

InChI

1S/C10H17NO2/c1-10(2,3)13-9(12)11-7-5-4-6-8-11/h5,7H,4,6,8H2,1-3H3

InChI key

VIBQTJLMJANION-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

199.9 °F - closed cup

Flash Point(C)

93.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service