Skip to Content
Merck
All Photos(1)

Key Documents

714941

Sigma-Aldrich

Chloro(cyclopentadienyl)[bis(diphenylphosphino)methane]ruthenium(II)

97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$232.00

$232.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$232.00

About This Item

Empirical Formula (Hill Notation):
C30H27ClP2Ru
CAS Number:
Molecular Weight:
586.01
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$232.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

190-196 °C

SMILES string

Cl[Ru].[CH]1[CH][CH][CH][CH]1.C(P(c2ccccc2)c3ccccc3)P(c4ccccc4)c5ccccc5

InChI

1S/C25H22P2.C5H5.ClH.Ru/c1-5-13-22(14-6-1)26(23-15-7-2-8-16-23)21-27(24-17-9-3-10-18-24)25-19-11-4-12-20-25;1-2-4-5-3-1;;/h1-20H,21H2;1-5H;1H;/q;;;+1/p-1

InChI key

NFGZGIRSDQBCFT-UHFFFAOYSA-M

Application

Catalyst for:
  • Anti-Markovnikov hydration
  • β-alkylation with primary alcohols
  • Cyclization of terminal alkynals to cycloalkenes
  • Electrochemical oxidation of methanol
  • Hydration of 1-alkynes
  • Isomerization of allylic alcohols

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service