Skip to Content
Merck
All Photos(2)

Key Documents

712922

Sigma-Aldrich

(S)-(+)-Naproxen chloride

≥97.0%

Synonym(s):

(S)-2-(6-Methoxy-2-naphthyl)propionyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$336.00

$336.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$336.00

About This Item

Empirical Formula (Hill Notation):
C14H13ClO2
CAS Number:
Molecular Weight:
248.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$336.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)
≥97.0%

form

crystals

optical activity

[α]/D +67.0±3.0°, c = 1 in methylene chloride

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(Cl)=O

InChI

1S/C14H13ClO2/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3/t9-/m0/s1

InChI key

UODROGXCIVAQDJ-VIFPVBQESA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service