Skip to Content
Merck
All Photos(1)

Key Documents

712752

Sigma-Aldrich

6-Azido-6-deoxy-D-galactose

≥98.0% (HPLC)

Synonym(s):

6-Azido-6-deoxygalactose

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$455.00

$455.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$455.00

About This Item

Empirical Formula (Hill Notation):
C6H11N3O5
CAS Number:
Molecular Weight:
205.17
Beilstein:
7569066
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$455.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 57.0±4.0°, c = 1 in H2O

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

OC1O[C@H](CN=[N+]=[N-])[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H11N3O5/c7-9-8-1-2-3(10)4(11)5(12)6(13)14-2/h2-6,10-13H,1H2/t2-,3+,4+,5-,6?/m1/s1

InChI key

CMLRUUHRGSJVMD-SVZMEOIVSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service