Skip to Content
Merck
All Photos(1)

Key Documents

712116

Sigma-Aldrich

meso-2,2′-Bipyrrolidine sesquioxalate salt

98%

Synonym(s):

(R*,S*)-2,2′-Bipyrrolidine sesquioxalate salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
MXP 1,560.00
500 G
MXP 5,407.00

MXP 1,560.00


Availability

In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
MXP 1,560.00
500 G
MXP 5,407.00

About This Item

Empirical Formula (Hill Notation):
C8H16N2 · C3H3O6
CAS Number:
Molecular Weight:
275.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

MXP 1,560.00


Availability

In StockDetails


Request a Bulk Order

Assay

≥98.5% (GC)
98%

form

powder

impurities

≤0.5% water

SMILES string

OC(=O)C(O)=O.C1CN[C@H](C1)[C@@H]2CCCN2.C3CN[C@H](C3)[C@@H]4CCCN4

InChI

1S/2C8H16N2.C2H2O4/c2*1-3-7(9-5-1)8-4-2-6-10-8;3-1(4)2(5)6/h2*7-10H,1-6H2;(H,3,4)(H,5,6)/t2*7-,8+;

InChI key

VSBQBHKEKBYWIP-BMROQPAASA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service