Skip to Content
Merck
All Photos(1)

Documents

711527

Sigma-Aldrich

5-Hydroxy-1-methyl-3-(trifluoromethyl)pyrazole

≥97%

Synonym(s):

1-Methyl-3-trifluoromethyl-1H-pyrazol-5-ol, 2-Methyl-5-trifluoromethyl-2H-pyrazol-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5F3N2O
CAS Number:
Molecular Weight:
166.10
Beilstein:
3542117
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97%

form

crystals

mp

172-178 °C

SMILES string

Cn1nc(cc1O)C(F)(F)F

InChI

1S/C5H5F3N2O/c1-10-4(11)2-3(9-10)5(6,7)8/h2,11H,1H3

InChI key

UGVAQFVQIHNCJV-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service