Skip to Content
Merck
All Photos(1)

Key Documents

711187

Sigma-Aldrich

5-Aminolevulinic acid-13C5, 15N hydrochloride

99 atom % 13C, 98 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$2,880.00

$2,880.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$2,880.00

About This Item

Linear Formula:
H215N13CH213CO(13CH2)213CO2H · HCl
Molecular Weight:
173.55
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.12

$2,880.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

Assay

97% (CP)

form

powder

technique(s)

bio NMR: suitable

mp

156 °C (dec.)

mass shift

M+6

storage temp.

−20°C

SMILES string

Cl.[15NH2][13CH2][13C](=O)[13CH2][13CH2][13C](O)=O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H/i1+1,2+1,3+1,4+1,5+1,6+1;

InChI key

ZLHFONARZHCSET-BVNCJLROSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Jericó et al.
Molecular therapy. Nucleic acids, 25, 207-219 (2021-08-31)
Variegate porphyria (VP) results from haploinsufficiency of protoporphyrinogen oxidase (PPOX), the seventh enzyme in the heme synthesis pathway. There is no VP model that recapitulates the clinical manifestations of acute attacks. Combined administrations of 2-allyl-2-isopropylacetamide and rifampicin in rabbits halved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service