Skip to Content
Merck
All Photos(1)

Key Documents

709573

Sigma-Aldrich

Desmethylraclopride

Desmethylraclopride
1 of 1 reviewers received a sample product or took part in a promotion

98% (CP)

Synonym(s):

Desmethylraclopride, 3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide, O-Desmethylraclopride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$885.00
25 MG
$3,270.00

$885.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
5 MG
$885.00
25 MG
$3,270.00

About This Item

Empirical Formula (Hill Notation):
C14H18Cl2N2O3
Molecular Weight:
333.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

$885.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Assay

98% (CP)

form

powder

technique(s)

PET imaging: suitable

storage temp.

−20°C

SMILES string

CCN1CCC[C@H]1CNC(=O)c2c(O)c(Cl)cc(Cl)c2O

InChI

1S/C14H18Cl2N2O3/c1-2-18-5-3-4-8(18)7-17-14(21)11-12(19)9(15)6-10(16)13(11)20/h6,8,19-20H,2-5,7H2,1H3,(H,17,21)/t8-/m0/s1

InChI key

DPQOMEPZWBXAMA-QMMMGPOBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service