Skip to Content

End of year dates: Orders must be billed by 13/12. Merck Argentina will be closed from 23/12 to 02/01.

Merck
All Photos(1)

Key Documents

70796

Sigma-Aldrich

(R)-3-(Fmoc-amino)-4-(2-naphthyl)butyric acid

≥95.0%

Synonym(s):

Fmoc-4-(2-naphthyl)-D-β-homoalanine, Fmoc-D-β-2-Homonal-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H25NO4
CAS Number:
Molecular Weight:
451.51
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95.0%

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)C[C@@H](Cc1ccc2ccccc2c1)NC(=O)OCC3c4ccccc4-c5ccccc35

InChI

1S/C29H25NO4/c31-28(32)17-22(16-19-13-14-20-7-1-2-8-21(20)15-19)30-29(33)34-18-27-25-11-5-3-9-23(25)24-10-4-6-12-26(24)27/h1-15,22,27H,16-18H2,(H,30,33)(H,31,32)/t22-/m1/s1

InChI key

LQGDRWACMZMSCD-JOCHJYFZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service