Skip to Content
Merck
All Photos(2)

Key Documents

704938

Sigma-Aldrich

N-Benzyl-2-bromo-N-phenylpropionamide

97%

Synonym(s):

2-Bromo-N-phenyl-N-(phenylmethyl)propanamide, N-Benzyl-2-bromo-propionanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16BrNO
CAS Number:
Molecular Weight:
318.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

75-80 °C

functional group

amide
bromo
phenyl

SMILES string

CC(Br)C(=O)N(Cc1ccccc1)c2ccccc2

InChI

1S/C16H16BrNO/c1-13(17)16(19)18(15-10-6-3-7-11-15)12-14-8-4-2-5-9-14/h2-11,13H,12H2,1H3

InChI key

SVPMVGLFGUEUOK-UHFFFAOYSA-N

Application

Used in Ni-pybox catalyzed asymmetric Negishi couplings.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Fischer et al.
Journal of the American Chemical Society, 127(13), 4594-4595 (2005-03-31)
A Ni/Pybox catalyst achieves the asymmetric cross-coupling of secondary alpha-bromo amides with organozinc reagents. The process tolerates a variety of functional groups and affords the desired product in good yield and in high enantiomeric excess.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service