Skip to Content
Merck
All Photos(1)

Key Documents

703761

Sigma-Aldrich

3,5-Diphenyl-4-(1-naphthyl)-1H-1,2,4-triazole

97% (HPLC)

Synonym(s):

TAZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17N3
CAS Number:
Molecular Weight:
347.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

solid

mp

242-246 °C

λmax

264 nm

fluorescence

λem 367 nm in dichloromethane-d2

Orbital energy

HOMO 6.4 eV 
LUMO 2.8 eV 

SMILES string

c1ccc(cc1)-c2nnc(-c3ccccc3)n2-c4cccc5ccccc45

InChI

1S/C24H17N3/c1-3-11-19(12-4-1)23-25-26-24(20-13-5-2-6-14-20)27(23)22-17-9-15-18-10-7-8-16-21(18)22/h1-17H

InChI key

AOQKGYRILLEVJV-UHFFFAOYSA-N

Application

Electron Transport Layer (ETL) material

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Color-variable highly efficient organic electrophosphorescent diodes manipulating molecular exciton and excimer emissions
Cocchi, M.; Kalinowski, J.; Fattori, V.; et al.
Applied Physics Letters, 94, 073309-073309 (2009)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service