Skip to Content
Merck
All Photos(1)

Key Documents

702927

Sigma-Aldrich

4-(Hydroxymethyl)phenylboronic acid pinacol ester

97%

Synonym(s):

(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 2-[4-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol, [4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$75.40
5 G
$181.00

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$75.40
5 G
$181.00

About This Item

Empirical Formula (Hill Notation):
C13H19BO3
CAS Number:
Molecular Weight:
234.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

74-80 °C

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(CO)cc2

InChI

1S/C13H19BO3/c1-12(2)13(3,4)17-14(16-12)11-7-5-10(9-15)6-8-11/h5-8,15H,9H2,1-4H3

InChI key

GZZBZWITJNATOD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Jun Wan et al.
International journal of pharmaceutics, 566, 731-744 (2019-06-19)
Tumor cells avoid immunosurveillance during the tumorigenesis, metastasis and recurrence periods thanks to the overexpressed immunosuppressive molecules on their surface. For instance, the programmed cell death 1 ligand (PD-L1) binds with the T-cells' programmed cell death receptor 1 (PD-1) impairing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service