Skip to Content
Merck
All Photos(2)

Key Documents

702412

Sigma-Aldrich

N-Boc-trans-4-fluoro-L-proline methyl ester

97%

Synonym(s):

1-tert-Butyl 2-methyl (2S,4R)-4-fluoro-1,2-pyrrolidinedicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$532.00

$532.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$532.00

About This Item

Empirical Formula (Hill Notation):
C11H18FNO4
CAS Number:
Molecular Weight:
247.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$532.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

97%

form

powder

optical activity

[α]22/D -80.0°, c = 1 in methanol

mp

34-39 °C

functional group

ester
fluoro

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H]1C[C@@H](F)CN1C(=O)OC(C)(C)C

InChI

1S/C11H18FNO4/c1-11(2,3)17-10(15)13-6-7(12)5-8(13)9(14)16-4/h7-8H,5-6H2,1-4H3/t7-,8+/m1/s1

InChI key

METPQQHVRNLTRX-SFYZADRCSA-N

Application

N-Boc-trans-4-fluoro-L-proline methyl ester can be used as a substrate in the preparation of pyrrolidinine-thioxotetrahydropyrimidinone based catalysts, which find application in asymmetric organic reactions.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Fluoro and 4-hydroxy pyrrolidine-thioxotetrahydropyrimidinones: Organocatalysts for green asymmetric transformations in brine
Kaplaneris N, et al.
The Journal of Organic Chemistry, 80(11), 5464-5473 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service