Skip to Content
Merck
All Photos(5)

Key Documents

702277

Sigma-Aldrich

Lithium titanate, spinel

greener alternative

nanopowder, <200 nm particle size (BET), >99%

Synonym(s):

LTO nanopowder, Lithium titanate spinel oxide, Lithium titanium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li4Ti5O12
CAS Number:
Molecular Weight:
459.09
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

>99%

form

nanopowder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

32.6 m2/g , BET

particle size

<200 nm (BET)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Li+].[Li+].[Li+].[O-][Ti](=O)O[Ti](O[Ti]([O-])=O)(O[Ti]([O-])=O)O[Ti]([O-])=O

InChI

1S/4Li.12O.5Ti/q4*+1;;;;;;;;;4*-1;;;;;

InChI key

BNQVSKURWGZJMY-UHFFFAOYSA-N

General description

Lithium titanate, spinel is an electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.[1][2][3][4]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium titanate, spinel nanopowder can be used as an anode material, which shows an ion conductivity of 10-13 Scm-1 at room temperature. It can also be used as an alternative to conventional graphite materials. It can further be used in the fabrication of high-performance lithium-ion batteries for electric vehicles (EVs).[5][6][7]

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Template-free synthesis of mesoporous spinel lithium titanate microspheres and their application in high-rate lithium ion batteries
Tang Y, et al.
Journal of Materials Chemistry, 19(33), 5980-5984 (2009)
Mesoporous spinel Li4Ti5O12 nanoparticles for high rate lithium-ion battery anodes
Liu W, et al.
Electrochimica Acta, 133, 578-582 (2014)
Sungmook Jung et al.
Scientific reports, 5, 17081-17081 (2015-11-26)
Wearable devices have attracted great attentions as next-generation electronic devices. For the comfortable, portable, and easy-to-use system platform in wearable electronics, a key requirement is to replace conventional bulky and rigid energy devices into thin and deformable ones accompanying the
Seongjun Bae et al.
ACS applied materials & interfaces, 7(30), 16565-16572 (2015-07-15)
Despite the many efforts to solve the problem associated with lithium storage at high rates, it is rarely achieved up until now. The design with experimental proof is reported here for the high rate of lithium storage via a core-shell
Synthesis and characterization of atomic layer deposited titanium nitride thin films on lithium titanate spinel powder as a lithium-ion battery anode
Snyder MQ, et al.
Journal of Power Sources, 165(1), 379-385 (2007)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service