Skip to Content
Merck
All Photos(1)

Key Documents

701769

Sigma-Aldrich

Dichloro(mesitylene)ruthenium(II) dimer

95%

Synonym(s):

(Mesitylene)dichlororuthenium dimer, Bis[dichloro(mesityl)ruthenium], Tetrachlorobis(mesitylene)diruthenium;, [RuCl2(mesitylene)]2, Di-μ-chlorodichlorobis[(1,2,3,4,5,6-η)-1,3,5-trimethylbenzene]diruthenium, Ruthenium(II) chloride mesitylene dimer

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$169.00

$169.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$169.00

About This Item

Empirical Formula (Hill Notation):
C18H24Cl4Ru2
CAS Number:
Molecular Weight:
584.34
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$169.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

crystals

reaction suitability

core: ruthenium
reagent type: catalyst

mp

>300 °C

storage temp.

2-8°C

SMILES string

Cl[Ru]1[Cl][Ru](Cl)[Cl]1.C[C]2[CH][C](C)[CH][C](C)[CH]2.C[C]3[CH][C](C)[CH][C](C)[CH]3

InChI

1S/2C9H12.2ClH.2Cl.2Ru/c2*1-7-4-8(2)6-9(3)5-7;;;;;;/h2*4-6H,1-3H3;2*1H;;;;/q;;;;;;2*+1/p-2

InChI key

SQSMPOQPOXFKBI-UHFFFAOYSA-L

Application

Catalyst for asymmetric hydrogenation, transfer hydrogenation, Diels-Alder reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, D. L.; Fawcett, J. et al.
Chemical Communications (Cambridge, England), 3151-3151 (1997)
Damjan Sterk et al.
Organic letters, 8(26), 5935-5938 (2006-12-15)
[Structure: see text] The asymmetric transfer hydrogenation of fluoroalkyl ketones mediated by [Ru(eta6-arene)((S,S)-R2NSO2DPEN)] catalysts using HCO2H-Et3N afforded the corresponding alcohols with high ee's and in excellent yields.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service