Skip to Content
Merck
All Photos(2)

Documents

700894

Sigma-Aldrich

4-Fluorobenzylsulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClFO2S
CAS Number:
Molecular Weight:
208.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

60-61 °C

SMILES string

Fc1ccc(CS(Cl)(=O)=O)cc1

InChI

1S/C7H6ClFO2S/c8-12(10,11)5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

UUQGWVIRPCRTSA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

225.0 °F

Flash Point(C)

107.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service